Orders LinkContactUsLinkComponent
L-Cysteic acid monohydrate ≥99% | Avantor
L-Cysteic acid monohydrate ≥99%
Codice catalogo
L-Cysteic acid monohydrate ≥99%
Codice catalogo
Nome del fornitore:  
Numero CAS:  23537-25-9
L-Cysteic acid monohydrate ≥99%
Numero CAS:  23537-25-9
Product Details & Documents
L-Cysteic Acid, Monohydrate, also known simply as cysteic acid or as 3-Sulfo-L-alanine, is used as an intermediate in cysteine metabolism.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Sinonimi:  2-Amino-3-sulphopropionic acid monohydrate
  • Numero CAS:23537-25-9
  • Formula:C₃H₇NO₅S·H₂O
  • Numero MDL:MFCD00149544
  • Molecular Weight:187.19 g/mol (169.17 anhy)
  • temperatura di conservazione:Ambiente
Specifiche
Risultati dei test sulle specifiche
  • Assay:
    Approx. 99%