Order Entry
Switzerland
ContactUsLinkComponent
Anti-C1orf84 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-C1orf84 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-15077R-A555
 :  Bioss
Anti-C1orf84 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-15077R-A555
 :  Bioss

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    C1orf84
  • Clonalità:
    Policlonale
  • Coniugazione:
    Alexa Fluor® 555
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Epitopo:
    651-750/3432
  • Forma:
    Liquid
  • ID gene:
    23334
  • Sinomini antigene:
    Hypothetical protein LOC149469|Uncharacterized protein C1orf84|Chromosome 1 open reading frame 84|SZT2_HUMAN
  • Modifica:
    Unmodified
  • Tampone di conservazione:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Peso molecolare:
    378 kDa
  • Temperatura di conservazione:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentrazione:
    1 µg/µl
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human C1orf84
  • Applicazioni testate:
    ICC
  • Purificazione:
    Purified by Protein A
  • Tipo di conf.:
    Vial
  • Conf:
    100 µl

 

 

SZT2 (seizure threshold 2 homolog (mouse)), also known as SZT2A or SZT2B, is a 3,432 amino acid peroxisomal protein that plays a role in resistance to oxidative stress. Predominantly expressed in the parietal and frontal cortex, as well as in dorsal root ganglia of the brain, SZT2 is implicated in superoxide dismutase activity and the neuroprotection in peroxisomes. Existing as four alternatively spliced isoforms, SZT2 is thought to enhance epileptogenesis and is encoded by a gene that maps to human chromosome 1p34.2. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson?s disease, Gaucher disease, schizophrenia and Usher syndrome.

Type: Primary
Antigen: C1orf84
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 651-750/3432
Host: Rabbit
Isotype: IgG
Reactivity: