Order Entry
Switzerland
ContactUsLinkComponent
Anti-LRRC6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-LRRC6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13613R-A680
Supplier:  Bioss
Anti-LRRC6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13613R-A680
Supplier:  Bioss

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    LRRC6
  • Clonalità:
    Policlonale
  • Coniugazione:
    Alexa Fluor® 680
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitopo:
    201-300/466
  • Forma:
    Liquid
  • Sinomini antigene:
    Protein TILB homolog| Leucine-rich repeat-containing protein 6| Leucine rich repeat containing protein 6| LRTP| TSLRP| Testis specic leucine rich repeat protein| TILB_HUMAN| Leucine-rich testis-specic protein| Leucine rich testis specic protein| Lrrc6| Testis-specic leucine-rich repeat protein|Leucine rich repeat containing 6
  • Modifica:
    Unmodified
  • Tampone di conservazione:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Peso molecolare:
    54 kDa
  • Temperatura di conservazione:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentrazione:
    1 µg/µl
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human LRRC6
  • Applicazioni testate:
    ICC
  • Purificazione:
    Purified by Protein A
  • Tipo di conf.:
    Vial
  • Conf:
    100 µl

Specifications

About this item

The leucine-rich repeat-containing protein 6 (LRRC6), also designated Leucine-rich testis-specific protein (LRTP), is a 466 amino acid protein that contains 3 LRR repeats and plays a role in spermatogenesis. The gene encoding LRRC6 maps to chromosome 8, which encodes approximately 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Chromosome 8 is also associated with Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.

Type: Primary
Antigen: LRRC6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/466
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse