Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-SNF5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6109R-A750
 :  Bioss
Anti-SNF5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-6109R-A750
 :  Bioss

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    SNF5
  • Clonalità:
    Policlonale
  • Coniugazione:
    Alexa Fluor® 750
  • Citometria a flusso:
    Yes
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Mouse
  • Epitopo:
    301-385/385
  • Forma:
    Liquid
  • ID gene:
    6598
  • Sinomini antigene:
    SNF5|hSNF5|SWI/SNF complex component SNF5|TYE4|Transcription regulatory protein SNF5|SNF5L1|Snr1|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1|RDT|Transcription factor TYE4|INI1 antibody Integrase interactor 1 protein|Sfh1p|SWI10|BAF47|SMARCB1|Sucrose nonfermenting yeast homolog like 1
  • Modifica:
    Unmodified
  • Tampone di conservazione:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Peso molecolare:
    44 kDa
  • Temperatura di conservazione:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentrazione:
    1 µg/µl
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human SNF5
  • Applicazioni testate:
    ICC
  • Purificazione:
    Purified by Protein A
  • Tipo di conf.:
    Vial
  • Conf:
    100 µl

 

 

SNF5 is involved in transcriptional activation. The SWI/SNF complex is required for the induced expression of a large number of genes. This complex alters chromatin structure to facilitate binding of gene-specific dedicated transcription factors. SNF5 also funtions as a tumor suppressor gene and is frequently mutated in malignant rhabdoid tumors. It is also involved in adipocyte differentiation.

Type: Primary
Antigen: SNF5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-385/385
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse