Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CNG channel beta 1/truncated GARP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-11332R-A750
 :  Bioss
Anti-CNG channel beta 1/truncated GARP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-11332R-A750
 :  Bioss

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    CNG channel beta 1/truncated GARP
  • Clonalità:
    Policlonale
  • Coniugazione:
    Alexa Fluor® 750
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Epitopo:
    851-950/1251
  • Forma:
    Liquid
  • ID gene:
    1258
  • Sinomini antigene:
    GARP|RP45|CNGB1|Cyclic nucleotide-gated cation channel gamma|Cyclic nucleotide-gated cation channel modulatory subunit|CNCG2|RCNCb|GAR1|GARP2|CNGB1B|CNG channel beta-1|CNG-4|CNCG3L|Glutamic acid-rich protein|CNG channel 4|RCNC2|Cyclic nucleotide-gated channel beta-1|CNG4|RCNCbeta|Cyclic nucleotide-gated cation channel 4|Cyclic nucleotide-gated cation channel beta-1|CNCG4
  • Modifica:
    Unmodified
  • Tampone di conservazione:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Peso molecolare:
    140 kDa
  • Temperatura di conservazione:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentrazione:
    1 µg/µl
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human truncated GARP
  • Applicazioni testate:
    ICC
  • Purificazione:
    Purified by Protein A
  • Tipo di conf.:
    Vial
  • Conf:
    100 µl

 

 

Subunit of cyclic nucleotide-gated (CNG) channels, nonselective cation channels, which play important roles in both visual and olfactory signal transduction. When associated with CNGA1, it is involved in the regulation of ion flow into the rod photoreceptor outer segment (ROS), in response to light-induced alteration of the levels of intracellular cGMP. Isoform GARP2 is a high affinity rod photoreceptor phosphodiesterase (PDE6)-binding protein that modulates its catalytic properties: it is a regulator of spontaneous activation of rod PDE6, thereby serving to lower rod photoreceptor 'dark noise' and allowing these sensory cells to operate at the single photon detection limit.

Type: Primary
Antigen: CNG channel beta 1/truncated GARP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 851-950/1251
Host: Rabbit
Isotype: IgG
Reactivity: