Order Entry
Switzerland
ContactUsLinkComponent
Anti-GPR113 Rabbit Polyclonal Antibody (Cy5)
Anti-GPR113 Rabbit Polyclonal Antibody (Cy5)
Catalog # BOSSBS-15375R-CY5
Supplier:  Bioss
Anti-GPR113 Rabbit Polyclonal Antibody (Cy5)
Catalog # BOSSBS-15375R-CY5
Supplier:  Bioss

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    GPR113
  • Clonalità:
    Policlonale
  • Coniugazione:
    Cy5®
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    1-100/1079
  • Forma:
    Liquid
  • Sinomini antigene:
    Probable G-protein coupled receptor 113|FLJ16767|G-protein coupled receptor PGR23|GPR113|PGR23|hGPCR37|GP113_HUMAN|UNQ9196/PRO34000|GP113
  • Modifica:
    Unmodified
  • Tampone di conservazione:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Peso molecolare:
    114 kDa
  • Temperatura di conservazione:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentrazione:
    1 µg/µl
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human GPR113
  • Applicazioni testate:
    ICC
  • Purificazione:
    Purified by Protein A
  • Tipo di conf.:
    Vial
  • Conf:
    100 µl

Specifications

About this item

GPR113.

Type: Primary
Antigen: GPR113
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 1-100/1079
Host: Rabbit
Isotype: IgG
Reactivity: Human