Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Codice catalogo 093050201.
Nome del fornitore: MP Biomedicals
Numero CAS:
Specifiche
- Conf:1 KIT
- Descrizione:Mycoplasma PCR kit
Specifiche
Informazioni su questo articolo
Myco-Sniff™ mycoplasma PCR detection kit provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
- Highly specific: No Interference of animal or bacterial DNA
- Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA
The detection limit for contaminating mycoplasma is as low as 20 CFU/ml. Includes 48 lyophilised PCR tubes for 20 μl reactions.