ContactUsLinkComponent
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric)
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric)
Codice catalogo ABCAAB283387-100
Nome del fornitore:  Abcam
Numero CAS:  
undefined
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric)
Codice catalogo ABCAAB283387-100
Nome del fornitore:  Abcam
Numero CAS:  
Specifiche
  • Conf:
    100 Tests
  • Descrizione:
    Carbonic anhydrase (CA) inhibitor screening kit (colorimetric), Size; 100 assays
Specifiche
Informazioni su questo articolo
It is simple and can be used to identify and characterize CA inhibitors in a high-throughput format.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of Carbonic Anhydrase ligands
  • Kit includes Acetazolamide, a CA inhibitor, and a stable, recombinant human CA
Carbonic anhydrases (CA; EC: 4.2.1.1) are zinc enzymes present in both prokaryotes and eukaryotes. They efficiently catalyse the reversible hydration of CO2 to bicarbonate. Their important patho-physiological roles in respiration, pH and CO2 homeostasis, secretion, gluconeogenesis, ureagenesis etc. make it an important drug target. CA inhibition in different organs can result in different effects. Inhibition of CA helps in different disease treatment procedures; for example, it can help in the treatment of Glaucoma, urine alkalisation, managing epilepsy, sleep apnea, acute mountain sickness etc. BioVision’s CA Inhibitor Screening Kit can be used to screen for potent inhibitors of CA activity. It utilises the esterase activity of an active CA on an ester substrate which releases a chromogenic product. The released product can be easily quantified using an absorbance microplate reader. In the presence of a CA specific inhibitor, the enzyme loses its activity which results in decrease of absorbance. This assay kit is simple and can be used to identify and characterize CA inhibitors in a high-throughput format. Here, we have used the specific inhibitor Acetazolamide, a potent inhibitor of CA I, CA II, CA IV, CA IX, CA XII etc. except CA III
Caution:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.