ContactUsLinkComponent
S-Glutathionylated Protein Detection Kit
S-Glutathionylated Protein Detection Kit
Codice catalogo CAYM10010721-1
Nome del fornitore:  Cayman Chemical
Numero CAS:  
undefined
S-Glutathionylated Protein Detection Kit
Codice catalogo CAYM10010721-1
Nome del fornitore:  Cayman Chemical
Numero CAS:  
Specifiche
  • Conf:
    1
  • Descrizione:
    Kit contains: Buffers and reagents
  • Nome marchio:
    CAYM
Specifiche
Informazioni su questo articolo
A convenient method for the direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells.
  • Direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells by flow cytometry and microscopy
  • Visualisation using colorimetric or fluorescence detection
  • Reagents provided to test up to 30 samples
Glutathione (GSH) is involved in maintenance of protein sulfhydryl reduction status. PSSG may modify the activity of a large number of cell proteins, including metabolic, structural, cytoskeletal, and signaling proteins. Cayman’s PSSG provides a convenient method for the direct visualisation of S-glutathionylated proteins in whole (permeabilised) cells by flow cytometry and microscopy as well as avidin overlay analysis. This cell-based assay starts with the modification of protein free-thiols groups followed by enzymatic cleavage of any PSSG adducts present in the sample. Biotinylation of the newly-formed protein free-thiols provides the basis for visualisation using streptavidin-based colorimetric or fluorescence detection. Reagents are provided to test three sets of 10 samples (most convenient) or up to thirty samples total at once if desired.