Quick-DNA™ Fecal/Soil Microbe Kits
Rapid methods for the isolation of inhibitor-free, PCR quality DNA from fecal, soil, and microbial samples in minutes including tough-to-lyse bacteria, fungi, algae, and protozoa.
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
- Zymo-Spin™ Column and unique filtration technologies effectively removes PCR inhibitors from the DNA product
The Quick-DNA™ Fecal/Soil Microbe Kits are designed for the simple and rapid isolation of inhibitor-free, PCR-quality DNA from microbes in soil and feces. This kit can be used to isolate DNA from variety of soils (clay, sandy, silty, peaty, chalky, and loamy) and feces (humans, birds, rats, mice, cattle and more). Tough-to-lyse bacteria, fungi, protozoa, and algae are all rapidly and efficiently lysed by bead beating with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ Column or Plate technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. The procedures can be performed in minutes, and there is no need for organic denaturants or proteinases. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection and more.
Rapid methods for the isolation of inhibitor-free, PCR quality DNA from fecal, soil, and microbial samples in minutes including tough-to-lyse bacteria, fungi, algae, and protozoa.
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
- Zymo-Spin™ Column and unique filtration technologies effectively removes PCR inhibitors from the DNA product
The Quick-DNA™ Fecal/Soil Microbe Kits are designed for the simple and rapid isolation of inhibitor-free, PCR-quality DNA from microbes in soil and feces. This kit can be used to isolate DNA from variety of soils (clay, sandy, silty, peaty, chalky, and loamy) and feces (humans, birds, rats, mice, cattle and more). Tough-to-lyse bacteria, fungi, protozoa, and algae are all rapidly and efficiently lysed by bead beating with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ Column or Plate technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. The procedures can be performed in minutes, and there is no need for organic denaturants or proteinases. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection and more.
- Catalog No:
- ZYMOD6011
- ZYMOD6012
- ZYMOD6110
- ZYMOD6010
- Descrizione:
- Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps
- Quick-DNA™ Fecal/soil microbe microprep kit, 50 preps
- Quick-DNA™ Fecal/soil microbe midiprep kit, 25 preps
- Quick-DNA™ Fecal/soil microbe miniprep kit, 50 preps
- Target molecule:
- DNA
- DNA
- DNA
- DNA
- Refine target molecule:
- Total DNA
- Total DNA
- Total DNA
- Total DNA
- Sample type:
- Stool|Soil
- Stool|Soil
- Stool|Soil
- Stool|Soil
- Method / Format:
- Multiwell plate
- Column (silica-based)
- Column (silica-based)
- Column (silica-based)
- Prep size:
- Mini
- Micro
- Midi
- Mini
Product Family Options
- DescrizioneFormatoConf
- Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps96 well spin column plate2 KIT1 * 2 KITSFr. 1’017.00Marketsource
Specifications
Cat. No.ZYMOD6011DescrizioneQuick-DNA™ Fecal/soil microbe 96 kit, 2× 96 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeStool|SoilMethod / FormatMultiwell platePrep sizeMiniMarketsource - Quick-DNA™ Fecal/soil microbe microprep kit, 50 prepsSpin column1 KIT1 * 1 KITSFr. 327.00Marketsource
Specifications
Cat. No.ZYMOD6012DescrizioneQuick-DNA™ Fecal/soil microbe microprep kit, 50 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeStool|SoilMethod / FormatColumn (silica-based)Prep sizeMicroMarketsource - Quick-DNA™ Fecal/soil microbe midiprep kit, 25 prepsSpin column1 KIT1 * 1 KITSFr. 718.00Marketsource
Specifications
Cat. No.ZYMOD6110DescrizioneQuick-DNA™ Fecal/soil microbe midiprep kit, 25 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeStool|SoilMethod / FormatColumn (silica-based)Prep sizeMidiMarketsource - Quick-DNA™ Fecal/soil microbe miniprep kit, 50 prepsSpin column1 KIT1 * 1 KITSFr. 327.00Marketsource
Specifications
Cat. No.ZYMOD6010DescrizioneQuick-DNA™ Fecal/soil microbe miniprep kit, 50 prepsTarget moleculeDNARefine target moleculeTotal DNASample typeStool|SoilMethod / FormatColumn (silica-based)Prep sizeMiniMarketsource



