Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Quick-DNA™ Plus Kits

Quick-DNA™ Plus Kits

Marketsource
Supplier:   Zymo Research
Quick-DNA™ Plus Kits
Supplier:  Zymo Research
Marketsource
Certificates
Quick-DNA™ Plus Kits

6 Options Available Below

Product Details & Documents

DNA from any sample (cells, tissues, biological fluids and more).

  • Quick and easy: Simple 20 minute procedure
  • Highest yield: Recover 3× more DNA
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA >50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ZYMOD4068T
  • ZYMOD4068
  • ZYMOD4069
  • ZYMOD4070
  • ZYMOD4071
  • ZYMOD4074
  • Descrizione:
  • Quick-DNA™ Miniprep Plus kit
  • Quick-DNA™ Miniprep Plus kit
  • Quick-DNA™ Miniprep Plus kit
  • Quick-DNA™ 96 Plus kit
  • Quick-DNA™ 96 Plus kit
  • Quick-DNA™ Microprep Plus kit
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • DNA
  • DNA
  • DNA
  • Refine target molecule:
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Sample type:
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Method / Format:
  • Column (silica-based)
  • Column (silica-based)
  • Column (silica-based)
  • Multiwell plate
  • Multiwell plate
  • Column (silica-based)
  • Prep size:
  • Mini
  • Mini
  • Mini
  • Mini
  • Mini
  • Mini
  • Dimensione del campione:
  • ≤5×10⁶ Cells or 25 mg Tissue
  • ≤5×10⁶ Cells or 25 mg Tissue
  • ≤5×10⁶ Cells or 25 mg Tissue
  • ≤1×10⁶ Cells or 5 mg Tissue
  • ≤1×10⁶ Cells or 5 mg Tissue
  • ≤1×10⁶ Cells or 5 mg Tissue
  • Capacità campioni:
  • 25 µg
  • 25 µg
  • 25 µg
  • 5 µg
  • 5 µg
  • 5 µg
  • Resa:
  • The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.

Specifications

Product Family Options

Product Information

  • Descrizione
    Volume
    Conf
    Availability
    Price
  • Quick-DNA™ Miniprep Plus kit
    10 Preps
    10 Tests
    Log in for availability
    Log in for availability
    1 * 10 Tests
    1 * 10 Tests - SFr. 65.00
    SFr. 65.00
    1 * 10 Tests
       SFr. 65.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4068T
    Descrizione
    Quick-DNA™ Miniprep Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Dimensione del campione
    ≤5×10⁶ Cells or 25 mg Tissue
    Capacità campioni
    25 µg
    Resa
    The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
  • Quick-DNA™ Miniprep Plus kit
    50 Preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 202.00
    SFr. 202.00
    1 * 1 KIT
       SFr. 202.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4068
    Descrizione
    Quick-DNA™ Miniprep Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Dimensione del campione
    ≤5×10⁶ Cells or 25 mg Tissue
    Capacità campioni
    25 µg
    Resa
    The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
  • Quick-DNA™ Miniprep Plus kit
    200 Preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 746.00
    SFr. 746.00
    1 * 1 KIT
       SFr. 746.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4069
    Descrizione
    Quick-DNA™ Miniprep Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Dimensione del campione
    ≤5×10⁶ Cells or 25 mg Tissue
    Capacità campioni
    25 µg
    Resa
    The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
  • Quick-DNA™ 96 Plus kit
    2×96 Preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 808.00
    SFr. 808.00
    1 * 1 KIT
       SFr. 808.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4070
    Descrizione
    Quick-DNA™ 96 Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Multiwell plate
    Prep size
    Mini
    Dimensione del campione
    ≤1×10⁶ Cells or 5 mg Tissue
    Capacità campioni
    5 µg
    Resa
    The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
  • Quick-DNA™ 96 Plus kit
    4×96 Preps
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’508.00
    SFr. 1’508.00
    1 * 1 KIT
       SFr. 1’508.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4071
    Descrizione
    Quick-DNA™ 96 Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Multiwell plate
    Prep size
    Mini
    Dimensione del campione
    ≤1×10⁶ Cells or 5 mg Tissue
    Capacità campioni
    5 µg
    Resa
    The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
  • Quick-DNA™ Microprep Plus kit
    50 Preps
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - SFr. 207.00
    SFr. 207.00
    1 * 50 Tests
       SFr. 207.00

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ZYMOD4074
    Descrizione
    Quick-DNA™ Microprep Plus kit
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
    Method / Format
    Column (silica-based)
    Prep size
    Mini
    Dimensione del campione
    ≤1×10⁶ Cells or 5 mg Tissue
    Capacità campioni
    5 µg
    Resa
    The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.