ContactUsLinkComponent
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
Catalog # HRDYW570
Supplier:  HARDY DIAGNOSTICS
CAS Number:  
undefined
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
Catalog # HRDYW570
Supplier:  HARDY DIAGNOSTICS
CAS Number:  
Specifications
  • Conf:
    1
  • Descrizione:
    SterEM™ Tryptic soy agar (TSA), irradiated, USP, triple bagged, 15×100 mm Plate, 26 ml
Specifications
About this item
Sterile plates for environmental monitoring.
  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation - Validated for Sterility Assurance Level (SAL) of 10-5
  • Triple bagged, two nylon inner bags and one outer Mylar(R) bag that is heat sealed
Irradiated Tryptic Soy Agar (TSA), USP is recommended for use as a general growth medium for the detection and enumeration of microorganisms. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Certified to ISO 13485 - Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.