Thromboxane B₂ ELISA Kit
For the quantification of TXB2 in urine, serum, tissue culture supernatants, and other sample matrices.
- Measure TXB2 in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure TXB2 levels down to 5 pg/ml
- Incubation: 18 hours | Development: 90 to 120 minutes | Read: Colorimetric at 405 to 420 nm
Thromboxane A2 (TXA2) is produced from arachidonic acid by many cells and causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. Like most lipid mediators, TXA2 is not a circulating hormone. It is formed in response to local stimuli and exerts its effects within a short distance of its biosynthesis. TXA2 is rapidly hydrolyzed non-enzymatically to form TXB2, which is then quickly metabolized (t½ = 5-7 minutes) to urinary metabolites for clearance by the kidneys. Because of the transient nature of this compound it is difficult to accurately measure circulating levels in whole animal experimental models. In fact, it has been shown that plasma and urine levels of TXB2 are primarily due to ex vivo platelet activation and intra-renal production, respectively. Therefore, measurement of TXB2 metabolites, such as 11-dehydro TXB2, in urine and plasma may give more accurate estimates of in vivo TXA2 production. TXB2 measurement is better suited for samples that are unlikely to undergo extensive metabolism such as perfusates, lavage samples, tissue/cell culture, etc.
: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate(s).
For the quantification of TXB2 in urine, serum, tissue culture supernatants, and other sample matrices.
- Measure TXB2 in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure TXB2 levels down to 5 pg/ml
- Incubation: 18 hours | Development: 90 to 120 minutes | Read: Colorimetric at 405 to 420 nm
Thromboxane A2 (TXA2) is produced from arachidonic acid by many cells and causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. Like most lipid mediators, TXA2 is not a circulating hormone. It is formed in response to local stimuli and exerts its effects within a short distance of its biosynthesis. TXA2 is rapidly hydrolyzed non-enzymatically to form TXB2, which is then quickly metabolized (t½ = 5-7 minutes) to urinary metabolites for clearance by the kidneys. Because of the transient nature of this compound it is difficult to accurately measure circulating levels in whole animal experimental models. In fact, it has been shown that plasma and urine levels of TXB2 are primarily due to ex vivo platelet activation and intra-renal production, respectively. Therefore, measurement of TXB2 metabolites, such as 11-dehydro TXB2, in urine and plasma may give more accurate estimates of in vivo TXA2 production. TXB2 measurement is better suited for samples that are unlikely to undergo extensive metabolism such as perfusates, lavage samples, tissue/cell culture, etc.
: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, and Plate(s).
- Catalog No:
- CAYM501020-480
- CAYM501020-96
- Target protein:
- Thromboxane B2 (TXB₂)
- Thromboxane B2 (TXB₂)
- Tipo di test:
- Competitive
- Competitive
- Formato:
- Pre-coated
- Pre-coated
- Durata del test:
- Multiple steps
- Multiple steps
- Sensibilità:
- 80% B/B0: 5 pg/ml
- 80% B/B0: 5 pg/ml
- Range di rilevamento:
- 1,6 - 1000 pg/ml
- 1,6 - 1000 pg/ml
- Metodo di rilevamento:
- Colorimetric at 405 - 420 nm
- Colorimetric at 405 - 420 nm
- Tiipo del campione:
- Urine, serum, tissue culture supernatants, and other sample matrices
- Urine, serum, tissue culture supernatants, and other sample matrices
- Tempo di sviluppo:
- 90 - 120 min
- 90 - 120 min
- Tempo di incubazione:
- 18 hr
- 18 hr
Product Family Options
- DescrizioneConf
- Thromboxane B₂ ELISA kit, 480 wells480 Tests1 * 480 TestsSFr. 2’780.00
Specifications
Cat. No.CAYM501020-480Target proteinThromboxane B2 (TXB₂)Tipo di testCompetitiveFormatoPre-coatedDurata del testMultiple stepsSensibilità80% B/B0: 5 pg/mlRange di rilevamento1,6 - 1000 pg/mlMetodo di rilevamentoColorimetric at 405 - 420 nmTiipo del campioneUrine, serum, tissue culture supernatants, and other sample matricesTempo di sviluppo90 - 120 minTempo di incubazione18 hr - Thromboxane B₂ ELISA kit, 96 wells96 Tests1 * 96 TestsSFr. 742.00
Specifications
Cat. No.CAYM501020-96Target proteinThromboxane B2 (TXB₂)Tipo di testCompetitiveFormatoPre-coatedDurata del testMultiple stepsSensibilità80% B/B0: 5 pg/mlRange di rilevamento1,6 - 1000 pg/mlMetodo di rilevamentoColorimetric at 405 - 420 nmTiipo del campioneUrine, serum, tissue culture supernatants, and other sample matricesTempo di sviluppo90 - 120 minTempo di incubazione18 hr