Zymo-Spin™ IIC Columns
Informazioni su questo articolo
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 50 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IIC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥25 μl eluate. Capacity is 900 μl.
2 varianti
Product Details & Documents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 50 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IIC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥25 μl eluate. Capacity is 900 μl.
Product Family Options
Informazioni sul prodotto
- FormatoConfDisponibilitàPrice
- ZYMOC1011-50Zymo-Spin™ IICSpecifiche:MarketsourceMarketsource
- ZYMOC1011-250Zymo-Spin™ IICSpecifiche:MarketsourceMarketsource