Zymo-Spin™ I Columns
Informazioni su questo articolo
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 5 µg
- RNA binding capacity: 10 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ I column can be used either in micro centrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.
2 varianti
Product Details & Documents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 5 µg
- RNA binding capacity: 10 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ I column can be used either in micro centrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.
Product Family Options
Informazioni sul prodotto
- FormatoConfDisponibilitàPrice
- ZYMOC1003-250Zymo-Spin™ ISpecifiche:MarketsourceMarketsource
- ZYMOC1003-50Zymo-Spin™ ISpecifiche:MarketsourceMarketsource