Orders LinkContactUsLinkComponent
Zymo-Spin™ I Columns
Marketsource
Nome del fornitore:   Zymo Research
Zymo-Spin™ I Columns
Nome del fornitore:  Zymo Research
Marketsource
Certificates
Zymo-Spin™ I Columns
Zymo-Spin™ I Columns
2 varianti
Product Details & Documents
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 5 µg
  • RNA binding capacity: 10 µg
  • Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ I column can be used either in micro centrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ I features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.
Product Family Options
Informazioni sul prodotto
  • Formato
    Conf
    Disponibilità
    Price
  • ZYMOC1003-250
    Zymo-Spin™ I
    Zymo-Spin™ I
    Spin Column
    250
    Log in per disponibilità
    Log in per disponibilità
    1 * 250 ST
    1 * 250 ST - SFr. 308.00
    SFr. 308.00
    1 * 250 ST
       SFr. 308.00
    Specifiche:
    Marketsource
    Marketsource
  • ZYMOC1003-50
    Zymo-Spin™ I
    Zymo-Spin™ I
    Spin Column
    50
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 ST
    1 * 50 ST - SFr. 73.00
    SFr. 73.00
    1 * 50 ST
       SFr. 73.00
    Specifiche:
    Marketsource
    Marketsource