Shield™ DNA/RNA Reagent
Informazioni su questo articolo
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.
- Nucleic acid preservation (at ambient temperature; cold-free)
- Pathogen inactivation (bacteria, fungus, parasites & viruses)
- RNA Stability: Ambient temperature (4 to 25 °C) >1 month
- DNA Stability: Ambient temperature (4 to 25 °C) >2 years
- DNA and RNA: Frozen (<–20 °C): Indefinitely
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialised equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).
3 varianti
Product Details & Documents
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.
- Nucleic acid preservation (at ambient temperature; cold-free)
- Pathogen inactivation (bacteria, fungus, parasites & viruses)
- RNA Stability: Ambient temperature (4 to 25 °C) >1 month
- DNA Stability: Ambient temperature (4 to 25 °C) >2 years
- DNA and RNA: Frozen (<–20 °C): Indefinitely
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialised equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- ZYMOR1200-25
- ZYMOR1200-125
- ZYMOR1100-50
- Descrizione:
- DNA/RNA Shield™ (2X Concentrate)
- DNA/RNA Shield™ (2X Concentrate)
- DNA/RNA Shield™ (1X Concentrate)
- Target molecule:
- Copurification of DNA & RNA|DNA|RNA
- Copurification of DNA & RNA|DNA|RNA
- Copurification of DNA & RNA|DNA|RNA
- Refine target molecule:
- Total RNA|Total DNA
- Total RNA|Total DNA
- Total RNA|Total DNA
- Sample type:
- Stool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Stool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Stool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Method / Format:
- Chemical extraction & preservation
- Chemical extraction & preservation
- Chemical extraction & preservation
- Prep size:
- Scalable
- Scalable
- Scalable
Specifiche
Product Family Options
Informazioni sul prodotto
- VolumeConfDisponibilitàPrice
- ZYMOR1200-25DNA/RNA Shield™ (2X Concentrate)Specifiche:Marketsource
More
Specifications
Cat. No.ZYMOR1200-25DescrizioneDNA/RNA Shield™ (2X Concentrate)Target moleculeCopurification of DNA & RNA|DNA|RNARefine target moleculeTotal RNA|Total DNASample typeStool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissuesMethod / FormatChemical extraction & preservationPrep sizeScalableMarketsource - ZYMOR1200-125DNA/RNA Shield™ (2X Concentrate)Specifiche:Marketsource
More
Specifications
Cat. No.ZYMOR1200-125DescrizioneDNA/RNA Shield™ (2X Concentrate)Target moleculeCopurification of DNA & RNA|DNA|RNARefine target moleculeTotal RNA|Total DNASample typeStool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissuesMethod / FormatChemical extraction & preservationPrep sizeScalableMarketsource - ZYMOR1100-50DNA/RNA Shield™ (1X Concentrate)Specifiche:Marketsource
More
Specifications
Cat. No.ZYMOR1100-50DescrizioneDNA/RNA Shield™ (1X Concentrate)Target moleculeCopurification of DNA & RNA|DNA|RNARefine target moleculeTotal RNA|Total DNASample typeStool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissuesMethod / FormatChemical extraction & preservationPrep sizeScalableMarketsource



