Reusable Respirator Spectacle Kit
Catalog # MMMASK102
Supplier: 3M
Specifications
- Descrizione:Reusable respirator spectacle kit
- Taglia:Universal
- Conf:1 KIT
Specifications
About this item
Wear prescription lenses while using certain 3M™ Reusable Full Face Masks with the help of 3M™ Reusable Respirator Spectacle Kit. These spectacle frames attach to 3M™ Reusable Full Face Masks and can be adjusted.
- Spectacle kit for 3M™ Reusable Full Face Masks
- Permits wearer to use corrective lenses and to adjust them to a comfortable position
- One each per case
- Easy installation
- For occupational use only
Get the correct level of comfort and protection from protective equipment, while using prescription spectacles, with the 3M™ Reusable Respirator Spectacle kit. The kit allows use of corrective lenses and adjusts their position for a suitable fit inside 3M™ Full Face Respirators. Easy to install, the spectacle kit helps to ensure worker safety and compliance for those workers requiring prescription eyewear.