Modular Basket System
Nome del fornitore: ELMA SCHMIDBAUER
Informazioni su questo articolo
Modular basket system for Elmasonic P, Elmasonic Select and EASY series ultrasonic baths.
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Hole diameter bottom (mm): 11×15
Hole diameter sides (mm): 10×10
10 varianti
Product Details & Documents
Modular basket system for Elmasonic P, Elmasonic Select and EASY series ultrasonic baths.
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Hole diameter bottom (mm): 11×15
Hole diameter sides (mm): 10×10
Specifications for Product Family
Specifiche
- Catalog No:
- ELMA1117923
- ELMA1119922
- ELMA1108916
- ELMA1113045
- ELMA1113050
- ELMA1111838
- ELMA1113387
- ELMA1113110
- ELMA1113404
- ELMA1113072
- Modello:
- Type 10
- Type 20
- Type 30
- Type 40
- Type 60
- Type 80
- Type 100
- Type 150
- Type 180
- Type 300
- L×P×A:
- 155×70×35 mm
- 110×105×50 mm
- 195×105×50 mm
- 195×105×95 mm
- 255×105×95 mm
- 455×105×55 mm
- 250×195×90 mm
- 450×255×55 mm
- 280×255×135 mm
- 450×255×135 mm
Specifiche
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- ELMA1117923Modular basket system type 10, chamfer 20×20 mm, dimension: 155×70×35 mmModular basket system type 10, chamfer 20×20 mm, dimension: 155×70×35 mmLog in per disponibilità1 * 1 STSFr. 56.90Specifiche:
More
Specifications
Cat. No.ELMA1117923ModelloType 10L×P×A155×70×35 mm - ELMA1119922Modular basket system type 20, chamfer 20×20 mm, dimension: 110×105×50 mmModular basket system type 20, chamfer 20×20 mm, dimension: 110×105×50 mmLog in per disponibilità1 * 1 STSFr. 60.60Specifiche:
More
Specifications
Cat. No.ELMA1119922ModelloType 20L×P×A110×105×50 mm - ELMA1108916Modular basket system type 30, chamfer 20×20 mm, dimension: 195×105×50 mmModular basket system type 30, chamfer 20×20 mm, dimension: 195×105×50 mmLog in per disponibilità1 * 1 STSFr. 63.00Specifiche:
More
Specifications
Cat. No.ELMA1108916ModelloType 30L×P×A195×105×50 mm - ELMA1113045Modular basket system type 40, chamfer 20×20 mm, dimension: 195×105×95 mmModular basket system type 40, chamfer 20×20 mm, dimension: 195×105×95 mmLog in per disponibilità1 * 1 STSFr. 74.10Specifiche:
More
Specifications
Cat. No.ELMA1113045ModelloType 40L×P×A195×105×95 mm - ELMA1113050Modular basket system type 60, chamfer 20×20 mm, dimension: 255×105×95 mmModular basket system type 60, chamfer 20×20 mm, dimension: 255×105×95 mmLog in per disponibilità1 * 1 STSFr. 89.00Specifiche:
More
Specifications
Cat. No.ELMA1113050ModelloType 60L×P×A255×105×95 mm - ELMA1111838Modular basket system type 80, chamfer 20×20 mm, dimension: 455×105×55 mmModular basket system type 80, chamfer 20×20 mm, dimension: 455×105×55 mmLog in per disponibilità1 * 1 STSFr. 167.00Specifiche:
More
Specifications
Cat. No.ELMA1111838ModelloType 80L×P×A455×105×55 mm - ELMA1113387Modular basket system type 100, chamfer 25×25 mm, dimension: 250×195×90 mmModular basket system type 100, chamfer 25×25 mm, dimension: 250×195×90 mmLog in per disponibilità1 * 1 STSFr. 192.00Specifiche:
More
Specifications
Cat. No.ELMA1113387ModelloType 100L×P×A250×195×90 mm - ELMA1113110Modular basket system type 150, chamfer 25×25 mm, dimension: 450×255×55 mmModular basket system type 150, chamfer 25×25 mm, dimension: 450×255×55 mmLog in per disponibilità1 * 1 STSFr. 247.00Specifiche:
More
Specifications
Cat. No.ELMA1113110ModelloType 150L×P×A450×255×55 mm - ELMA1113404Modular basket system type 180, chamfer 25×25 mm, dimension: 280×255×135 mmModular basket system type 180, chamfer 25×25 mm, dimension: 280×255×135 mmLog in per disponibilità1 * 1 STSFr. 223.00Specifiche:
More
Specifications
Cat. No.ELMA1113404ModelloType 180L×P×A280×255×135 mm - ELMA1113072Modular basket system type 300, chamfer 25×25 mm, dimension: 450×255×135 mmModular basket system type 300, chamfer 25×25 mm, dimension: 450×255×135 mmLog in per disponibilità1 * 1 STSFr. 247.00Specifiche:
More
Specifications
Cat. No.ELMA1113072ModelloType 300L×P×A450×255×135 mm
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Cover for round basket, stainless steel, Ext.Ø×H: 64×2 mmCover for round basket, stainless steel, Ext.Ø×H: 64×2 mmELMA3000006018Log in per disponibilità1 * 1 ST - SFr. 17.30
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Round basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006013Log in per disponibilità1 * 1 ST - SFr. 44.50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Round basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006012Log in per disponibilità1 * 1 ST - SFr. 27.20
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Cover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mmCover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mmELMA3000006016Log in per disponibilità1 * 1 ST - SFr. 22.30
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Round basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006014Log in per disponibilità1 * 1 ST - SFr. 50.70
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Minature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mmMinature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mmELMA3000006017Log in per disponibilità1 * 1 ST - SFr. 17.30
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Round basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mmRound basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mmELMA3000009104Log in per disponibilità1 * 1 ST - SFr. 50.70
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Round basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006015Log in per disponibilità1 * 1 ST - SFr. 79.10
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Level set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mmLevel set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mmELMA1125227Log in per disponibilità1 * 1 ST - SFr. 40.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Level set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mmLevel set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mmELMA1125228Log in per disponibilità1 * 1 ST - SFr. 44.50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mmElma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mmELMA1125213Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mmElma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mmELMA1125209Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mmElma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mmELMA1125210Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mmElma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mmELMA1125216Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Dividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mmDividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mmELMA1125221Log in per disponibilità1 * 1 ST - SFr. 50.70
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)ELMA1124042Log in per disponibilità1 * 1 ST - SFr. 63.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Silicone holder set (10), Ext.Ø×H: 20×20 mmSilicone holder set (10), Ext.Ø×H: 20×20 mmELMA1125208Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Holder set, stainless steel
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Level set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mmLevel set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mmELMA1125226Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mmElma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mmELMA1125214Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mmElma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mmELMA1125215Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Dividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mmDividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mmELMA1125223Log in per disponibilità1 * 1 ST - SFr. 44.50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Silicone holder 10×bottom comb set, 105×10×35 mmSilicone holder 10×bottom comb set, 105×10×35 mmELMA1125217Log in per disponibilità1 * 1 ST - SFr. 76.60
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Dividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mmDividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mmELMA1125220Log in per disponibilità1 * 1 ST - SFr. 32.20
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Dividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mmDividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mmELMA1125222Log in per disponibilità1 * 1 ST - SFr. 44.50
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Dividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mmDividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mmELMA1125224Log in per disponibilità1 * 1 ST - SFr. 50.70
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Silicone holder 10×side comb set, 150×10×45 mmSilicone holder 10×side comb set, 150×10×45 mmELMA1125218Log in per disponibilità1 * 1 ST - SFr. 76.60
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mmElma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mmELMA1125212Log in per disponibilità1 * 1 ST - SFr. 63.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Elma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mmElma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mmELMA1125211Log in per disponibilità1 * 1 ST - SFr. 19.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Level set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mmLevel set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mmELMA1125225Log in per disponibilità1 * 1 ST - SFr. 22.30