Orders LinkContactUsLinkComponent
Cleanroom Tunics
Cleanroom Tunics
Cleanroom Tunics
Cleanroom Tunics
Cleanroom Tunics
These shirts are made of Movix (96% polyester, 4% carbon). Suitable for C and D classes or ISO 7 to 9 cleanrooms. For higher cleanroom classes worn as an undergarment piece under a coverall. Can also be used for clean areas.
  • V-neck
  • Long inserted sleeves
  • Seamed at waist
Usable in ISO 7 to 9.
 : IEST-RP-CC003.5
  • Catalog No:
  • ALSO1033607-XS
  • ALSO1033607-S
  • ALSO1033607-M
  • ALSO1033607-L
  • ALSO1033607-XL
  • ALSO1033607-XXL
  • ALSO1033607-3XL
  • Applicazioni per abiti:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Genere:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Tessuto:
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Peso:
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
Product Family Options
  • Colore
    Taglia dell'abbigliamento
  • blu scuro
    XS
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-XS
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    S
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-S
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    M
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-M
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    L
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-L
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    XL
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-XL
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    XXL
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-XXL
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²
  • blu scuro
    3XL
    1 * 1 ST
    1 * 1 ST - 40.20 SFr.
    40.20 SFr.
    1 * 1 ST
       40.20 SFr.
    Specifications
    Cat. No.
    ALSO1033607-3XL
    Applicazioni per abiti
    Cleanroom
    Risk area
    Anti-static
    Genere
    Unisex
    Tessuto
    Movix anti-static (96% polyester, 4% carbon)
    Peso
    152 g/m²