Orders LinkContactUsLinkComponent
Oligo Pro II System
Oligo Pro II System
Oligo Pro II system, High throughput automated capillary electrophoresis instrument for purity analysis of oligonucleotides, compatible with the Oligo Pro II capillary arrays
Codice catalogo AGLSM5340AA
Nome del fornitore:   AGILENT
Oligo Pro II System
Codice catalogo AGLSM5340AA
Nome del fornitore:  AGILENT
Oligo Pro II System
Oligo Pro II system, High throughput automated capillary electrophoresis instrument for purity analysis of oligonucleotides, compatible with the Oligo Pro II capillary arrays
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Oligonucleotide purity analysis using parallel capillary electrophoresis with UV detection.
  • Fast separations - complete 12, 24, or 96 samples in about 1 hour
  • Improve product quality - monitor synthesis performance to demonstrate purity of ssDNA or ssRNA
  • Wide sizing range - achieve single base resolution through 60-mers
  • Throughput for all labs - variable throughput with the option of 12, 24, or 96 capillary arrays
  • Direct detection of oligos - no dye labeling needed, uses UV detection
  • Simplified report generation - advanced peak integration software and data reporting capabilities
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
The Agilent Oligo Pro II system delivers accurate purity assessments of ssDNA and ssRNA oligonucleotides without the use of dyes. Ensure optimum downstream performance of oligonucleotides up to 60-mer in length with n-1 resolution.
With 12-, 24-, and 96-capillary arrays available along with three 96-well plate sample drawers, the Oligo Pro II system quickly adjusts to meet changing throughput demands. Intuitive and automated operation simplify the measurement of oligonucleotide purity and help you focus on results.
Caution: For research use only. Not for use in diagnostic procedures.
Specifiche
  • Descrizione:
    Oligo Pro II system
  • Details:
    High throughput automated capillary electrophoresis instrument for purity analysis of oligonucleotides, compatible with the Oligo Pro II capillary arrays
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Details
    Disponibilità
    Price
  • Waste tray
    Waste tray
    AGLSP60-22NM
    For use with the Oligo Pro II system
    Log in per disponibilità
    1 * 1 ST - SFr. 29.65