Orders LinkContactUsLinkComponent
ZAG DNA Analyser System
ZAG DNA Analyser System
ZAG DNA analyser system, High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
Codice catalogo 732-4439
Nome del fornitore:   AGILENT
ZAG DNA Analyser System
Codice catalogo 732-4439
Nome del fornitore:  AGILENT
ZAG DNA Analyser System
ZAG DNA Analyser System
ZAG DNA analyser system, High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Parallel capillary electrophoresis system for automated high-throughput DNA fragment analysis.
  • High separation resolution - reliably distinguish and size DNA fragments within 3 bp of each other
  • High throughput - process 48, 96-well sample trays in 24 hours. Broad sizing range - size fragments between 35 bp and 20,000 bp through a variety of available DNA analysis kits
  • Fast separation times - samples can be separated in as little as 20 minutes
  • Intuitive data analysis - analytical software can process separation data automatically after the run and generate a report in multiple formats
  • desktop PC supplied with a comprehensive 3-year warranty through HP
The Agilent ZAG DNA analyser system relieves bottlenecks in automated high-throughput DNA fragment analysis workflows. Using parallel capillary electrophoresis, the ZAG DNA Analyser system can process thousands of DNA fragment samples a day, eliminating bottlenecks associated with agarose gels.
This system is ideal for a range of DNA fragment applications, from basic PCR fragment analysis to microsatellite detection. Minimal sample preparation, automated separation of samples, and the option for continuous operation help keep your workflow moving.
Attenzione: For research use only. Not for use in diagnostic procedures.
Specifiche
  • Descrizione:
    ZAG DNA analyser system
  • Details:
    High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Details
    Disponibilità
    Price
  • 96 well trays for buffer and waste locations, 50 per pack
    96 well trays for buffer and waste locations, 50 per pack
    AGLSP60-20
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in per disponibilità
    1 * 50 ST - SFr. 566.47
Informazioni sul prodotto
  • Codice catalogo
    Details
    Disponibilità
    Price
  • Large open waste tray/tub
    Large open waste tray/tub
    AGLSP60-22
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in per disponibilità
    1 * 1 ST - SFr. 29.65
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • ZAG DNA analyser verification kit, for the ZAG DNA analyser system
    ZAG DNA analyser verification kit, for the ZAG DNA analyser system
    AGLSZAG-OPQ