Pierce™ Firefly Luc One-Step Glow Assay Kit
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.
- Sensitive—highly sensitive detection of firefly luciferase activity
- Odorless—reagents have no foul or toxic odor, eliminating the need to work in a fume hood
- Convenient—one-step, mix-and-read protocol; no additional reagents required
- Stable—increased signal half-life and stability compared to flash assays
- Simple—does not require a luminometer with injectors
- Automation-friendly—amenable to high throughput assays
Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.
Delivery: Kit includes Firefly Luc One-Step Substrate, lyophilised (makes 1 µl of 1X); Firefly Luc One-Step Assay Buffer (11 ml).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifications
- Condizioni di conservazione:–20 °C