- Esterno L×P×A:747×769×1818 mm
- Capacità:499/289 L (gross/net)
- Descrizione:Freestanding freezer with SmartFrost, FFFsg 5501 Performance
- Seq:03
- Modello:FFFsg 5501
- Elettrico:~220 - 240 V, 50 Hz, 2,0 A
- Refrigerante:R 290
- Allarme:Visual and audible
- Scongelamento:Manual
- Display:Monochrome external digital display, touch buttons
- Blocco sportello:Mechanical
- Tipo sportello:Steel, self closing
- Ripiani/cassetti:Adjustable, wire shelves plastic-coated, 10 baskets
- Peso:99 (gross)/72 (net) kg
- Isolamento:63/67 mm
- Conf:1
- Interno LxPxA:580×523×1388 mm
- Temperatura:−9...−26 °C (ambient 10…40 °C)
Freezers designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooling power. They are enormously robust, long-lasting, easy to clean and particularly sustainable, not only thanks to natural refrigerant. At the same time, they fulfil all the valid directives, standards and norms, including in terms of hygiene and HACCP concept. The efficient cooling system not only ensures low energy and overall operating cost, also safe storage of items not requiring validation of temperature stability and homogeneity.
- Freezer range of –9 to –26 °C, Class L1 with SmartFrost
- Static cooling system
- Power failure alarm
- LAN/WiFi optional (SmartModule) and Local AP optional
- 75 cm wide models: Euronorm 600×400 mm (crosswise insertion)
- Internal liner material made of commercial-grade moulded polystyrol
Appliances with solid door: Climate class 5, self-closing door at <90° and mechanical lock, aluminium door handle.
Features: Individually adjustable maintenance timer. Castors/adjustable feet as accessory, 75 cm wide appliances with adjustable feet as standard. Steel/white external cabinet finish.
Climate class (energy directive): 5, Climate class (safety standard): 7, Temperature class (energy directive): L1.
75 cm wide models internal dimensions: 600×400 mm (600 mm wide), 75 cm wide models include a full compliment of wire baskets as standard.
Height of adjustable feet: 150 to 180 mm.