Disposable conductive booties
Disposable conductive booties, blu
Codice catalogo 114-9321
Nome del fornitore: DESCO
Vai a
Product Details & Documents
Disposable conductive surgical style Booties are 'one size fits all' with elastic opening.
- Lightweight
- Cleanroom compatible
- Maintains grounding with full mobility
- Will not scratch or mark flooring
Disposable ESD Booties can be used for person/footwear/flooring system per EN 61340-5-1.
Packaging: Sold 150 pairs per box.
Specifiche
- Colore:blu
- Descrizione:Disposable conductive booties
- Cat. No.:114-9321
- Antistatico:+