Printer Paper
Nome del fornitore: AGILENT HEWLETT PACKARD
Vai a
Informazioni su questo articolo
5 varianti
Product Details & Documents
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- HEWL5181-1219Perforated paper, 8,5×11" rolls, inkjet for Agilent 3397A, 3396 series II, and 3396A/B/95/94Perforated paper, 8,5×11" rolls, inkjet for Agilent 3397A, 3396 series II, and 3396A/B/95/94Log in per disponibilità1 * 4 ROU
- HEWL5181-1255Perforated paper, A4 size rolls, inkjet, for Agilent 3397A, 3396 series III, 3396A and B, 3395, 3394Perforated paper, A4 size rolls, inkjet, for Agilent 3397A, 3396 series III, 3396A and B, 3395, 3394Log in per disponibilità1 * 4 ROU
- HEWL5080-8800Thermal paper, blue print, for Agilent 3390/92/93
- HEWL5062-3561Z-fold paper, 8,5×11" rolls, for Agilent 3397A, 3396 series II, and 3396A/B/95/94Z-fold paper, 8,5×11" rolls, for Agilent 3397A, 3396 series II, and 3396A/B/95/94Log in per disponibilità1 * 5 ST
- HEWL9270-0658Thermal Z-fold paper, black print, 8,5×11", for Agilent 338X/5830/3370Thermal Z-fold paper, black print, 8,5×11", for Agilent 338X/5830/3370Log in per disponibilità1 * 8 ST