UDI adapters (1-96), sparQ
Catalog # 733-2897
Supplier: Quantabio
CAS Number:
Specifications
- Conf:1 KIT
- Descrizione:sparQ UDI adapters (1-96)
Specifications
About this item
sparQ UDI Adapters (1-96) are unique dual-indexed adapters designed to be used on illumina sequencing platforms. It is compatible with both DNA and RNA libraries. The use of UDI’s minimises index hopping events while also enabling greater multiplexing on flow cells.
- Flexible pooling: Multiplex up to 96 samples per sequencing run
- Improved performance: Prevents index hopping and enhances demultiplexing accuracy
- Optimised: For maximum ligation efficiency with sparQ library prep kits
- Multiple applications: Including whole genome (with amplification or PCR-free), target enrichment, whole transcriptome sequencing and many more