Portable steps with safety handle
Codice catalogo TEKMST00010/2
Nome del fornitore: TEKNOMEK
Specifiche
- Descrizione:Portable steps with safety handle, non-slip matting
- Seq:02
- L×P×A:535×380×1120 mm
- Conf:1
Specifiche
Informazioni su questo articolo
Our portable steps unit is 2 steps, fully portable strong and reliable workmate.
- Nonslip steps
- Brushed dull polish finish
- 2 step unit
- Fully portable
- Available in a checker plate version
Our portable steps unit is 2 steps, fully portable strong and reliable workmate. Fully welded in 304 grade stainless steel, choose from non-slip matting steps or steps with check plate and non-shedding surface. Moreover, it comes with a brushed dull polish finish making it hygienic and easy for you to clean.