Storage cupboards with internal cabinet
Catalog # TEKMSHCBIC12X60X17
Supplier: TEKNOMEK
Specifications
- Descrizione:Storage cupboard with internal cabinet
- Materiale:Acciaio inossidabile 304
- Seq:04
- L×P×A:1200×600×1780 mm
- Conf:1
Specifications
About this item
Our storage cupboard comes complete with an internal lockable cabinet.
- Brushed dull polish finish
- Internal adjustable shelf
- Stainless steel adjustable feet with non-marking rubber base
- Hinged lockable doors
- Internal lockable drug storage safe
Our storage cupboard is ideal for storing pharmaceuticals safely. An internal lockable cabinet and hinged lockable external doors ensure security, while an adjustable shelf provides practical storage.
Ordering information: Available in two different depths and widths.