96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
Catalog # 734-4137
Supplier: Corning
Specifications
- N. di pozzetti:96
- Bordo:Semi-skirted
- Colore del pozzetto:trasparente
- Descrizione:PCR plate, with 2 barcode labels, Thermowell® GOLD
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:I-RECs, ISO 14001 - Reynosa
- Con bar code:Yes
- Conf:50
Specifications
About this item
These half skirted 96 well PCR microplates feature a rigid construction that is ideal for automation.
- Clear medical grade PP is suitable for RT-PCR
- Uniform wall thinness assures efficient heat transfer while minimizing well-to-well variability
- 100% tested for well integrity and DNase- and RNase-free
- Nonsterile, without lids
- With two generic bar code labels per plate