Fentanyl resistant nitrile exam glove with extended cuff.
- Tested against both fentanyl and gastric acid (vomit) to simulate extreme, real world overdose situations
- Easy to see when hands have come into contact with potentially hazardous powders due to the dark, black color
- Provides extra protection over the wrist and forearm with an extended cuff
- Made with a soft, flexible nitrile formulation
- Fully textured for a reliable, consistent grip
- Certified to the Emergency Medical Glove Requirements of NFPA 1999
MICROFLEX MidKnight XTRA gloves feature a robust design and extended cuff, for protection in rugged environments. Their dark color helps mask the appearance of dirt, grease, and grime. This means wearing MICROFLEX MidKnight XTRA disposable gloves also makes it easy to see potentially hazardous light-colored substances and potential contaminations.
Proven to resist fentanyl for up to 240 minutes, these black disposable gloves meet the National Fire Protection Association (NFPA 1999) emergency medical operations Standard on Protective Clothing and Ensembles. This makes MICROFLEX MidKnight XTRA the ideal NFPA certified gloves for emergency medical professionals, law enforcement officers, maintenance workers, and others at risk of accidental exposure to fentanyl. MICROFLEX MidKnight XTRA gloves are made of a soft yet strong nitrile, for comfortable, durable protection in the event of chemical splashes. Fully textured, they help wearers confidently grip tools and materials. These NFPA certified gloves have an extended cuff, for added protection over the wrist and forearm. They are medical exam grade certified for use by healthcare professionals.
Food contact approved
- Catalog No:
- ANSE93-862/8.5-9
- ANSE93-862/7.5-8
- ANSE93-862/10.5-11
- ANSE93-862/6.5-7
- ANSE93-862/9.5-10
- Ambidestro/mano specifico:
- ambidestro
- ambidestro
- ambidestro
- ambidestro
- ambidestro
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- Lattice/senza lattice:
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Materiale:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finitura:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Tipo polsino:
- Esteso
- Esteso
- Esteso
- Esteso
- Esteso
- Spessore delle dita:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- Spessore palmo:
- 0,15 mm
- 0,15 mm
- 0,15 mm
- 0,15 mm
- 0,15 mm
- Regolamentato da FDA:
- +
- +
- +
- +
- +
- Norma EN:
- EN 374-1:2016 Type B (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (1-4)
- EN 374-1:2016 Type B (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (1-4)
- EN 374-1:2016 Type B (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (1-4)
- EN 374-1:2016 Type B (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (1-4)
- EN 374-1:2016 Type B (JKPT), EN ISO 374-5 VIRUS, EN 421, EN 455 (1-4)
- Sterilità:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Antistatico:
- +
- +
- +
- +
- +
- Approvati per il contatto con gli alimenti:
- +
- +
- +
- +
- +