Cleanroom hoods, CleanMax®
Codice catalogo 114-9543
Nome del fornitore: LAKELAND CLEANMAX APPAREL
Specifiche
- Taglia dell'abbigliamento:One size
- Applicazioni per abiti:Cleanroom
- Colore:bianco
- Seq:01
- Conf:100 PAIR
- Classe di cleanroom:100/ISO 5
Specifiche
Informazioni su questo articolo
Clean manufactured disposable apparel are individually packaged and folded to promote easier gowning with less excursions.
- Clean manufactured in silicone-free environment
- Latex-free
- Tunnelled elastic on wrists with thumb loops
- Zipper closure
- For critical environments
- Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All apparel provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Packaging: 100/case, individually packed.