Accessories for Rotary Vacuum Concentrator, RVC 2-25 CDplus universal package
Codice catalogo CHRS110369
Nome del fornitore: CHRIST MARTIN
Specifiche
- Descrizione:Angular rotor made of aluminium, black anodized, tube dimensions Ø 12×36 mm with flat base, setting angle 40°
- Conf:1
- Per:Total capacity 3 rotors for a total of 108 vessels