Coliform PA broth dehydrated culture media
Catalog # HIMEM1051-500G
Anbieter: HIMEDIA
CAS-Nummer:
Technische Daten
- Conf:500 g
- Descrizione:Coliform PA broth
Technische Daten
Über diesen Artikel
For the determination of the presence or absence of coliform bacteria in examination of pollution in treated water from water treatment plants or distribution systems.
- Microbiological culture media
- 500 g poly bottles