EPA vial convenience kits, assembled
Catalog # NSCAB7921
Anbieter: THERMO NATIONAL SCIENTIFIC
Technische Daten
- Tappo incluso:Yes
- Capacità:20,00 ml
- Colore:marrone
- Descrizione:EPA vial convenience kit
- Seq:01
- Conf:100
Technische Daten
Über diesen Artikel
Thermo Scientific™ national EPA vial convenience kits are designed for discrete water sampling applications.
Kits come with caps, septa and a certificate of analysis.