Orders LinkContactUsLinkComponent
Ammonio Methacrylate Copolymer , Reference Standards
Ammonio Methacrylate Copolymer , Reference Standards
Nome del fornitore:   USP
Ammonio Methacrylate Copolymer , Reference Standards
Nome del fornitore:  USP
Certificates
Ammonio Methacrylate Copolymer , Reference Standards
2 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • 1029909.
  • 1029910.
  • Descrizione:
  • Ammonio Methacrylate Copolymer Type A
  • Ammonio Methacrylate Copolymer Type B
  • CAS No.:
  • 33434-24-1
  • 33434-24-1
  • Formula:
  • (C₉H₁₈NO₂.C₅H₈O₂.C₅H₈O₂.Cl)n
  • (C₉H₁₈NO₂.C₅H₈O₂.C₅H₈O₂.Cl)x
Specifiche
Product Family Options
Informazioni sul prodotto
  • Standard
    Tipo di conf.
    Conf
    Disponibilità
    Price
  • 1029909.
    Ammonio Methacrylate Copolymer Type A
    Ammonio Methacrylate Copolymer Type A
    USP reference standard
    Vial
    500 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 100 mg
    1 * 100 mg - SFr. 450.00
    SFr. 450.00
    1 * 100 mg
       SFr. 450.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1029909.
    Descrizione
    Ammonio Methacrylate Copolymer Type A
    CAS No.
    33434-24-1
    Formula
    (C₉H₁₈NO₂.C₅H₈O₂.C₅H₈O₂.Cl)n
  • 1029910.
    Ammonio Methacrylate Copolymer Type B
    Ammonio Methacrylate Copolymer Type B
    USP reference standard
    Vial
    500 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 500 mg
    1 * 500 mg - SFr. 470.00
    SFr. 470.00
    1 * 500 mg
       SFr. 470.00
    Specifiche:
    More
    Specifications
    Cat. No.
    1029910.
    Descrizione
    Ammonio Methacrylate Copolymer Type B
    CAS No.
    33434-24-1
    Formula
    (C₉H₁₈NO₂.C₅H₈O₂.C₅H₈O₂.Cl)x