Orders LinkContactUsLinkComponent
Rivastigmine Tartrate, Reference Standards
Rivastigmine Tartrate, Reference Standards
 :   USP
Rivastigmine Tartrate, Reference Standards
 :  USP
Rivastigmine Tartrate, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
  • Catalog No:
  • 1604847.
  • 1604858.
  • Descrizione:
  • Rivastigmine Tartrate R-Isomer
  • Rivastigmine Tartrate
  • Synonym(s):
  • (R)-3-[1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate, hydrogen tartrate
  • CAS No.:
  • 415973-05-6
  • 129101-54-8
  • Formula:
  • C₁₄H₂₂N₂O₂.C₄H₆O₆
  • C₁₈H₂₈N₂O₈
  • N. ONU:
  • 2811
  • 2811
Product Family Options
  • Descrizione
    Standard
    Tipo di conf.
    Conf
  • Rivastigmine Tartrate R-Isomer
    USP reference standard
    Vial
    20 mg
    1 * 20 mg
    1 * 20 mg - SFr. 1’370.00
    SFr. 1’370.00
    1 * 20 mg
       SFr. 1’370.00
    Specifications
    Cat. No.
    1604847.
    Descrizione
    Rivastigmine Tartrate R-Isomer
    Synonym(s)
    (R)-3-[1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate, hydrogen tartrate
    CAS No.
    415973-05-6
    Formula
    C₁₄H₂₂N₂O₂.C₄H₆O₆
    N. ONU
    2811
  • Rivastigmine Tartrate
    USP reference standard
    Vial
    200 mg
    1 * 200 mg
    1 * 200 mg - SFr. 1’060.00
    SFr. 1’060.00
    1 * 200 mg
       SFr. 1’060.00
    Specifications
    Cat. No.
    1604858.
    Descrizione
    Rivastigmine Tartrate
    Synonym(s)
    CAS No.
    129101-54-8
    Formula
    C₁₈H₂₈N₂O₈
    N. ONU
    2811