Naproxen Related Compounds, Reference Standard
Informazioni su questo articolo
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- 1457425.
- 1457312.
- 1457436.
- 1457414.
- Descrizione:
- Naproxen Related Compound L
- Naproxen Related Compound A
- Naproxen Related Compound G
- Naproxen Related Compound K
- Synonym(s):
- 1-(6-methoxynaphthalen-2-yl)ethanone
- 6-methoxy-2-naphthoic acid
- (R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid
- 1-(6-Methoxynaphthalen-2-yl)ethanol
- CAS No.:
- 3900-45-6
- 2471-70-7
- 23979-41-1
- 77301-42-9
- Formula:
- C₁₃H₁₂O₂
- C₁₂H₁₀O₃
- N. ONU:
- 2811
Specifiche
Product Family Options
Informazioni sul prodotto
- StandardTipo di conf.ConfDisponibilitàPrice
- 1457425.Naproxen Related Compound LNaproxen Related Compound LUSP reference standardVial15 mgLog in per disponibilità1 * 15 mgSFr. 1’410.00Specifiche:
More
Specifications
Cat. No.1457425.DescrizioneNaproxen Related Compound LSynonym(s)1-(6-methoxynaphthalen-2-yl)ethanoneCAS No.3900-45-6FormulaC₁₃H₁₂O₂N. ONU - 1457312.Naproxen Related Compound ANaproxen Related Compound AUSP reference standardVial20 mgLog in per disponibilità1 * 20 mgSFr. 1’440.00Specifiche:
More
Specifications
Cat. No.1457312.DescrizioneNaproxen Related Compound ASynonym(s)6-methoxy-2-naphthoic acidCAS No.2471-70-7FormulaC₁₂H₁₀O₃N. ONU - 1457436.Naproxen Related Compound GNaproxen Related Compound GUSP reference standardVial50 mgLog in per disponibilità1 * 50 mgSFr. 1’340.00Specifiche:
More
Specifications
Cat. No.1457436.DescrizioneNaproxen Related Compound GSynonym(s)(R)-2-(6-Methoxynaphthalen-2-yl)propanoic acidCAS No.23979-41-1FormulaN. ONU2811 - 1457414.Naproxen Related Compound KNaproxen Related Compound KUSP reference standardVial50 mgLog in per disponibilità1 * 50 mgSFr. 1’370.00Specifiche:
More
Specifications
Cat. No.1457414.DescrizioneNaproxen Related Compound KSynonym(s)1-(6-Methoxynaphthalen-2-yl)ethanolCAS No.77301-42-9FormulaN. ONU