Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Hydrocortisone Butyrate, Reference Standard

Hydrocortisone Butyrate, Reference Standard

Hydrocortisone Butyrate, USP reference standard, 200 mg
Codice catalogo 1317302.
Nome del fornitore:   USP
Hydrocortisone Butyrate, Reference Standard
Codice catalogo 1317302.
Nome del fornitore:  USP
Numero CAS:  
Hydrocortisone Butyrate, Reference Standard

Hydrocortisone Butyrate, USP reference standard, 200 mg

Numero CAS:  
Price
SFr. 403.00
Log in per disponibilità
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Conf:
    200 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Hydrocortisone Butyrate
  • Cat. No.:
    1317302.
  • Formula:
    C₂₅H₃₆O₆
  • Descrizione:
    Hydrocortisone Butyrate
  • CAS No.:
    13609-67-1

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.