Orders LinkContactUsLinkComponent
Heparin Sodium, Reference Standards
Heparin Sodium, Reference Standards
Supplier:   USP
Heparin Sodium, Reference Standards
Supplier:  USP
Certificates
5 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1304016.
  • 1304038.
  • 1304049.
  • 1304050.
  • 1304107.
  • Descrizione:
  • Heparin Sodium for Assays
  • Heparin Sodium Identification
  • Heparin Sodium System Suitability
  • Heparin Sodium with Oversulfated Chondroitin Sulfate
  • Heparin Sodium Molecular Weight Calibrant
  • CAS No.:
  • 9041-08-1
  • 9041-08-1
  • 9041-08-1
  • 9041-08-1
  • 9041-08-1
  • Formula:
  • C₉H₂₀N₃NaO₃
  • C₉H₂₀N₃NaO₃
  • C₉H₂₀N₃NaO₃
  • C₉H₂₀N₃NaO₃
Specifications
Product Family Options
Product Information
  • Descrizione
    Standard
    Tipo di conf.
    Conf
    Availability
    Price
  • Heparin Sodium for Assays
    USP reference standard
    Ampule
    9,5 mg
    Log in for availability
    Log in for availability
    1 * 9,5 mg
    1 * 9,5 mg - 495.00 SFr.
    495.00 SFr.
    1 * 9,5 mg
       495.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    1304016.
    Descrizione
    Heparin Sodium for Assays
    CAS No.
    9041-08-1
    Formula
    C₉H₂₀N₃NaO₃
  • Heparin Sodium Identification
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 495.00 SFr.
    495.00 SFr.
    1 * 50 mg
       495.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    1304038.
    Descrizione
    Heparin Sodium Identification
    CAS No.
    9041-08-1
    Formula
    C₉H₂₀N₃NaO₃
  • Heparin Sodium System Suitability
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 230.00 SFr.
    1 230.00 SFr.
    1 * 50 mg
       1 230.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    1304049.
    Descrizione
    Heparin Sodium System Suitability
    CAS No.
    9041-08-1
    Formula
    C₉H₂₀N₃NaO₃
  • Heparin Sodium with Oversulfated Chondroitin Sulfate
    USP reference standard
    Ampule
    8,8 mg
    Log in for availability
    Log in for availability
    1 * 8,8 mg
    1 * 8,8 mg - 1 330.00 SFr.
    1 330.00 SFr.
    1 * 8,8 mg
       1 330.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    1304050.
    Descrizione
    Heparin Sodium with Oversulfated Chondroitin Sulfate
    CAS No.
    9041-08-1
    Formula
  • Heparin Sodium Molecular Weight Calibrant
    USP reference standard
    Ampule
    10,3 mg
    Log in for availability
    Log in for availability
    1 * 10,3 mg
    1 * 10,3 mg - 1 610.00 SFr.
    1 610.00 SFr.
    1 * 10,3 mg
       1 610.00 SFr.
    Specifications:
    More
    Specifications
    Cat. No.
    1304107.
    Descrizione
    Heparin Sodium Molecular Weight Calibrant
    CAS No.
    9041-08-1
    Formula
    C₉H₂₀N₃NaO₃