ContactUsLinkComponent
Dipyridamole Related Compounds, Reference Standard
 :  USP
Dipyridamole Related Compounds, Reference Standard
 :  USP
  • CAS No.:
    54093-92-4
  • Synonym(s):
    2,2'-{[6-chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]azanediyl}diethanol
  • Descrizione:
    Dipyridamole Related Compound C
Frequently Bought Together