Chrysanthemum Indicum Flower Dry Extract, Reference Standard
1133740.
: USP
:
- Tipo di conf.:Vial
- Conf:500 mg
- Standard:USP reference standard
- Descrizione:Chrysanthemum Indicum Flower Dry Extract
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard