Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Phytonadione, Reference Standard

Phytonadione, Reference Standard

Phytonadione, USP reference standard, 100 mg x 5 ampules, Bottiglia di vetro, 1 SET
Codice catalogo 1538006.
Nome del fornitore:   USP
Phytonadione, Reference Standard
Codice catalogo 1538006.
Nome del fornitore:  USP
Numero CAS:  
Phytonadione, Reference Standard

Phytonadione, USP reference standard, 100 mg x 5 ampules, Bottiglia di vetro, 1 SET

Numero CAS:  
Price
SFr. 471.00
Log in per disponibilità
1 * 1 SET
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    1 SET
  • Standard:
    USP reference standard
  • Descrizione:
    Phytonadione
  • Pk Info:
    100 mg x 5 ampules
  • Cat. No.:
    1538006.
  • Descrizione:
    Phytonadione
  • Synonym(s):
    Vitamin K1
  • CAS No.:
    81818-54-4

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..