Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Enanthic Acid, Reference Standard

Enanthic Acid, Reference Standard

Enanthic Acid, USP reference standard, Ampule, 100 mg
Codice catalogo 1235398.
Nome del fornitore:   USP
Enanthic Acid, Reference Standard
Codice catalogo 1235398.
Nome del fornitore:  USP
Numero CAS:  
Enanthic Acid, Reference Standard

Enanthic Acid, USP reference standard, Ampule, 100 mg

Numero CAS:  
Price
SFr. 967.00
Log in per disponibilità
1 * 100 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Tipo di conf.:
    Ampule
  • Conf:
    100 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Enanthic Acid
  • Cat. No.:
    1235398.
  • N. ONU:
    3265
  • Descrizione:
    Enanthic Acid
  • Synonym(s):
    Heptanoic Acid
  • CAS No.:
    111-14-8

Specifiche