Castor Oil, Reference Standards
Informazioni su questo articolo
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Castor Oil: Mixture containing about 92% of Triricinolein
Certifications: US Pharmacopea Reference Standard
2 varianti
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Castor Oil: Mixture containing about 92% of Triricinolein
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- 1096826.
- 1096837.
- Descrizione:
- Castor Oil
- Hydrogenated Castor Oil
- CAS No.:
- 8001-79-4
- 8001-78-3
Specifiche
Product Family Options
Informazioni sul prodotto
- StandardPk InfoTipo di conf.ConfDisponibilitàPrice
- 1096826.Castor OilCastor OilUSP reference standard3 x 100 mgAmpule300 mgLog in per disponibilità1 * 300 mg419.00 SFr.Specifiche:
More
Specifications
Cat. No.1096826.DescrizioneCastor OilCAS No.8001-79-4 - 1096837.Hydrogenated Castor OilHydrogenated Castor OilUSP reference standardVial250 mgLog in per disponibilità1 * 250 mg437.00 SFr.Specifiche:
More
Specifications
Cat. No.1096837.DescrizioneHydrogenated Castor OilCAS No.8001-78-3