Abacavir System Suitability Mixtures, Reference Standard
Abacavir System Suitability Mixture, USP reference standard, 15 mg
Codice catalogo 1000500.
Nome del fornitore: USP
Abacavir System Suitability Mixtures, Reference Standard
Codice catalogo 1000500.
Nome del fornitore: USP
Numero CAS:
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
A mixture containing abacavir sulfate and trans-abacavir
: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Conf:15 mg
- Standard:USP reference standard
- Descrizione:Abacavir System Suitability Mixture