PFA tongs
PFA tongs
Codice catalogo SAVI700-850
Nome del fornitore: SAVILLEX
Vai a
Product Details & Documents
Adjustable PFA tongs are moulded from the highest purity PFA resin for ultratrace metals use.
Service temperature range is –200 to +260 °C (–328 to +500 °F).
Ideal for transferring ICP glassware during cleaning. Tongs are 28 cm (11") long with three adjustable width positions.
Specifiche
- Descrizione:PFA tongs