Orders LinkContactUsLinkComponent
Hand held tool and covers
Hand held tool and covers
Nome del fornitore:   MICRONOVA
Hand held tool and covers
Nome del fornitore:  MICRONOVA
Certificates
Hand held tool and covers
Hand held tool and covers
3 varianti
Product Details & Documents
The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
  • Ergonomic cleaning for small equipment and surfaces
  • Lightweight, forms to hand
  • Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
  • Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
  • PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
  • Covers compatible with alcohol and strong disinfectants and are launderable for reuse
  • Autoclavable
The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.
Product Family Options
Hand held mop adapter
Informazioni sul prodotto
  • L×L
    Materiale
    Disponibilità
    Price
  • MICRHHT-06PP
    Hand held tool
    Hand held tool
    152×102 mm
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 28.90
    SFr. 28.90
    1 * 1 ST
       SFr. 28.90
Mop cover
Informazioni sul prodotto
  • L×L
    Materiale
    Disponibilità
    Price
  • MICRMZSM6-6NS
    NovaPoly™ slip cover, 100% smooth knit polyester, no foam, irradiated
    NovaPoly™ slip cover, 100% smooth knit polyester, no foam, irradiated
    152×102 mm
    Poliestere
    Log in per disponibilità
    Log in per disponibilità
    1 * 24 ST
    1 * 24 ST - SFr. 601.00
    SFr. 601.00
    1 * 24 ST
       SFr. 601.00
  • MICRMZSM68-6S
    PolyMesh™ slip cover, 100% polyester with mesh, foam interior
    PolyMesh™ slip cover, 100% polyester with mesh, foam interior
    152×102 mm
    Poliestere
    Log in per disponibilità
    Log in per disponibilità
    1 * 24 ST
    1 * 24 ST - SFr. 654.00
    SFr. 654.00
    1 * 24 ST
       SFr. 654.00