Swinging bucket rotor assembly, SW 60 Ti, Optima
Catalog # 521-3688
Supplier: Beckman Coulter
Specifications
- Descrizione:SW 60 Ti swinging bucket rotor assembly. Includes: SW 60 Ti rotor bucket set
- Conf:1
- Per:Swinging bucket (ultracentrifugation)
- Max. RPM:60000
- No. of tubes accepted:8×5 ml
- Raggio:120,3
mm