VWR®, Absorption Cuvettes
High quality cuvettes manufactured from quartz or optical glass for accurate spectrophotometric analysis.
- Fully heat-fused construction
- Two cover types available: Flat lid and stopper
- Matched sets available
Due to the fully heat-fused construction (no intermediate materials or adhesives), and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells provide a more suitable seal for volatile liquids. Matched cells exhibit a transmission tolerance within 0,5% at 200 nm (quartz cuvette)/360 nm (optical glass).
: Manufactured under ISO 19001:2015. Certificate of Quality is attached with each product.
: Packs of two cuvettes are matched sets. For cuvettes used in fluorometers, please refer to 634-1223 series.
High quality cuvettes manufactured from quartz or optical glass for accurate spectrophotometric analysis.
- Fully heat-fused construction
- Two cover types available: Flat lid and stopper
- Matched sets available
Due to the fully heat-fused construction (no intermediate materials or adhesives), and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells provide a more suitable seal for volatile liquids. Matched cells exhibit a transmission tolerance within 0,5% at 200 nm (quartz cuvette)/360 nm (optical glass).
: Manufactured under ISO 19001:2015. Certificate of Quality is attached with each product.
: Packs of two cuvettes are matched sets. For cuvettes used in fluorometers, please refer to 634-1223 series.
Product Family Options
Flat PTFE lids
- DescrizioneTipoAltezzaVolumeCammino otticoMateriale finestra
- MacroRounded bottom45 mm350 µl1 mmQuartz glass1 * 1 STSFr. 101.00
- MacroRounded bottom45 mm700 µl2 mmQuartz glass1 * 1 STSFr. 101.00
- Semi-microFlat bottom45 mm1 ml10 mmQuartz glass1 * 1 STSFr. 145.00
- Semi-microFlat bottom45 mm1,4 ml10 mmQuartz glass1 * 1 STSFr. 111.00
- MacroRounded bottom45 mm1,75 ml5 mmQuartz glass1 * 1 STSFr. 102.00
- MacroRounded bottom45 mm3,5 ml10 mmQuartz glass1 * 1 STSFr. 72.10
- Macro, spectrally matchedRounded bottom45 mm3,5 ml10 mmQuartz glass1 * 2 STSFr. 173.00
- MacroRounded bottom45 mm3,5 ml10 mmOptical glass1 * 1 STSFr. 46.60
- Macro, spectrally matchedRounded bottom45 mm3,5 ml10 mmOptical glass1 * 2 STSFr. 117.00
- MacroFlat bottom45 mm3,5 ml10 mmOptical glass1 * 1 STSFr. 35.60
- MacroFlat bottom45 mm3,5 ml10 mmQuartz glass1 * 1 STSFr. 64.30
- MacroRounded bottom45 mm7 ml20 mmQuartz glass1 * 1 STSFr. 114.00
- MacroRounded bottom45 mm7 ml20 mmOptical glass1 * 1 STSFr. 62.20
- MacroFlat bottom45 mm7 ml20 mmOptical glass1 * 1 STSFr. 57.70
- Semi-microFlat bottom45 mm7 ml50 mmQuartz glass1 * 1 STSFr. 233.00
- MacroRounded bottom45 mm14 ml40 mmQuartz glass1 * 1 STSFr. 144.00
- MacroRounded bottom45 mm14 ml40 mmOptical glass1 * 1 STSFr. 80.80
- MacroRounded bottom45 mm17,5 ml50 mmQuartz glass1 * 1 STSFr. 159.00
- MacroRounded bottom45 mm17,5 ml50 mmOptical glass1 * 1 STSFr. 98.80
- MacroFlat bottom45 mm17,5 ml50 mmOptical glass1 * 1 STSFr. 79.80
- MacroRounded bottom45 mm35 ml100 mmQuartz glass1 * 1 STSFr. 285.00
- MacroRounded bottom45 mm35 ml100 mmOptical glass1 * 1 STSFr. 168.00
PTFE stoppers
- DescrizioneTipoAltezzaVolumeCammino otticoMateriale finestra
- MacroFlat bottom52 mm350 µl1 mmQuartz glass1 * 1 STSFr. 154.00
- MacroFlat bottom52 mm700 µl2 mmQuartz glass1 * 1 STSFr. 154.00
- MacroRounded bottom45 mm1,75 ml5 mmQuartz glass1 * 1 STSFr. 135.00
- MacroRounded bottom45 mm3,5 ml10 mmQuartz glass1 * 1 STSFr. 126.00
- MacroRounded bottom45 mm7 ml20 mmQuartz glass1 * 1 STSFr. 172.00