Orders LinkContactUsLinkComponent
Fixed-angle rotor package/assembly, TLA-100
Fixed-angle rotor package/assembly, TLA-100
Nome del fornitore:   Beckman Coulter
Fixed-angle rotor package/assembly, TLA-100
Nome del fornitore:  Beckman Coulter
Certificates
Fixed-angle rotor package/assembly, TLA-100
2 varianti
Product Details & Documents
Major applications: Pelleting of subcellular fractions; proteins; RNA.
Note: Non-precipitating solutions up to 1,7 g/ml in density can be run in this rotor without a reduction in rotor speed.
Specifications for Product Family
Specifiche
  • Catalog No:
  • 521-3759
  • 521-3760
  • Capacità:
  • 4,0 ml
  • 4,0 ml
  • No. of tubes accepted:
  • 20×0 ml
  • 20×0 ml
  • Raggio:
  • 38,9 mm
  • 38,9 mm
  • Max. RPM:
  • 100000
  • 100000
Specifiche
Product Family Options
Rotor package
Informazioni sul prodotto
  • Per
    Disponibilità
    Price
  • 521-3759
    TLA-100 fixed-angle rotor package, includes 30° fixed-angle rotor (titanium), 1 box (BECM343775) thickwall polycarbonate tubes, 1 each (BECM348302) tube rack, (BECM824412 and BECM824953) O-rings, (BECM927208) Hemostats, (BECM346133) rotor vise/stand, (BECM306812) Spinkote lubricant, and (BECM335148) vacuum grease
    TLA-100 fixed-angle rotor package, includes 30° fixed-angle rotor (titanium), 1 box (BECM343775) thickwall polycarbonate tubes, 1 each (BECM348302) tube rack, (BECM824412 and BECM824953) O-rings, (BECM927208) Hemostats, (BECM346133) rotor vise/stand, (BECM306812) Spinkote lubricant, and (BECM335148) vacuum grease
    Optima MAX-XP Benchtop Ultracentrifuge, Optima TLX personal micro-ultracentrifuges, and Optima MAX high-capacity personal micro-ultracentrifuges, and the discontinued TL-100 and; Optima TL micro-ultracentrifuges
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 23’939.00
    SFr. 23’939.00
    1 * 1 ST
       SFr. 23’939.00
    Specifiche:
    More
    Specifications
    Cat. No.
    521-3759
    Capacità
    4,0 ml
    No. of tubes accepted
    20×0 ml
    Raggio
    38,9 mm
    Max. RPM
    100000
Rotor assembly
Informazioni sul prodotto
  • Per
    Disponibilità
    Price
  • 521-3760
    TLA-100 rotor assembly, fixed-angle (30° angle), titanium
    TLA-100 rotor assembly, fixed-angle (30° angle), titanium
    Optima MAX-XP Benchtop Ultracentrifuge, Optima TLX personal micro-ultracentrifuges, and Optima MAX high-capacity personal micro-ultracentrifuges, and the discontinued TL-100 and; Optima™ TL micro-ultracentrifuges
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 23’096.00
    SFr. 23’096.00
    1 * 1 ST
       SFr. 23’096.00
    Specifiche:
    More
    Specifications
    Cat. No.
    521-3760
    Capacità
    4,0 ml
    No. of tubes accepted
    20×0 ml
    Raggio
    38,9 mm
    Max. RPM
    100000